4-Chloro-4'-methyl-2-(trifluoromethyl)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

27996-500MG 27996-1G 27996-5G 27996-25G
77324-918EA 199.82 USD
77324-918 77348-752 77361-244 77384-504
4-Chloro-4'-methyl-2-(trifluoromethyl)-1,1'-biphenyl ≥95%
4-Chloro-4'-methyl-2-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClF₃
MW: 270.677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28142632
CAS Number: 1419798-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR