2-Methyl-3-nitro-6-phenoxypyridine

Supplier: AOBChem USA

Warning

104795-500MG 104795-1G 104795-5G
77337-634EA 431.57 USD
77337-634 77347-440 77371-442
2-Methyl-3-nitro-6-phenoxypyridine
2-Methyl-3-nitro-6-phenoxypyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28991939
CAS Number: 28232-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR