GDC-0152 98+%

Supplier: Ambeed
Warning

A852996-10MG A852996-1MG A852996-50MG A852996-5MG
77244-528EA 57.68 USD
77244-528 77244-530 77244-532 77244-534
GDC-0152 98+%
GDC-0152

Precaución: for Research Use Only

Formula: C₂₅H₃₄N₆O₃S
MDL Number: MFCD26142660
CAS Number: 873652-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR