4-(Dimethylamino)-1-naphthaldehyde 95%
Supplier: Ambeed
|
Warning
|
A384280-100MG
A384280-1G
A384280-250MG
77291-014EA
15
USD
77291-014
77616-670
77748-775
4-(Dimethylamino)-1-naphthaldehyde 95%
4-(Dimethylamino)-1-naphthaldehyde
Precaución: for Research Use Only
Formula:
C₁₃H₁₃NO |
MDL Number:
MFCD00010371 CAS Number: 1971-81-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...