1-(4-Nitrophenyl)-1H-pyrrole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A955954-1G A955954-250MG A955954-5G
77200-402EA 53 USD
77200-402 77200-404 77200-406
1-(4-Nitrophenyl)-1H-pyrrole-2-carboxylic acid 97%
1-(4-Nitrophenyl)-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O₄
MW: 232.19 g/mol
MDL Number: MFCD00541956
CAS Number: 82552-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR