2-(3-Buten-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98% stabilised

Supplier: Ambeed
Warning

Synonyms: (3-Buten-1-yl)boronic acid Pinacol ester, 2-(But-3-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Stabilization: Stabilised with phenothiazine

A284661-1G A284661-250MG A284661-5G
77166-678EA 33 USD
77166-678 77166-680 77166-682
2-(3-Buten-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98% stabilised
2-(3-Buten-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₀H₁₉BO₂
MDL Number: MFCD11858602
CAS Number: 331958-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR