WHI-P131 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-((6,7-Dimethoxyquinazolin-4-yl)amino)phenol
A712945-100MG
A712945-10MG
A712945-1G
A712945-1MG
A712945-250MG
A712945-25MG
A712945-50MG
A712945-5MG
77158-552EA
99
USD
77158-552
77158-554
77158-556
77158-558
77158-560
77158-562
77158-564
77158-566
WHI-P131 98%
WHI-P131
Precaución: for Research Use Only
Formula:
C₁₆H₁₅N₃O₃ MW: 297.3086 g/mol |
MDL Number:
MFCD01862614 CAS Number: 202475-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...