WHI-P180 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-((6,7-Dimethoxyquinazolin-4-yl)amino)phenol
A162125-10MG
A162125-1MG
A162125-2MG
A162125-50MG
A162125-5MG
77138-196EA
40.17
USD
77138-196
77138-198
77138-200
77138-202
77138-204
WHI-P180 98%
WHI-P180
Precaución: for Research Use Only
Formula:
C₁₆H₁₅N₃O₃ MW: 297.31 g/mol |
MDL Number:
MFCD02683970 CAS Number: 211555-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...