1-[(tert-Butyl)oxycarbonyl]-4-phenylpyrrolidine-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
trans-1-(tert-Butoxycarbonyl)-4-phenylpyrrolidine-3-carboxylic acid
A676661-100MG
A676661-250MG
A676661-1G
77104-760EA
53
USD
77104-760
77104-762
77901-072
1-[(tert-Butyl)oxycarbonyl]-4-phenylpyrrolidine-3-carboxylic acid 98%
1-[(tert-Butyl)oxycarbonyl]-4-phenylpyrrolidine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₆H₂₁NO₄ |
MDL Number:
MFCD02089475 CAS Number: 221142-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...