2-(2-Pyridyl)benzo[b]thiophene 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Benzo[b]thiophen-2-yl)pyridine, 2-[Benzo[b]thiophen-2-yl]pyridine

A680851-100MG A680851-1G A680851-250MG A680851-5G
76992-582EA 35 USD
76992-582 76992-584 76992-586 76992-588
2-(2-Pyridyl)benzo[b]thiophene 98%
2-(2-Pyridyl)benzo[b]thiophene

Precaución: for Research Use Only

Formula: C₁₃H₉NS
MDL Number: MFCD08457652
CAS Number: 38210-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR