Ethyl-6-chloroimidazo[1,2-a]pyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

A153499-100MG A153499-1G
77010-914EA 10.61 USD
77010-914 77010-916
Ethyl-6-chloroimidazo[1,2-a]pyridine-3-carboxylate 97%
Ethyl 6-chloroimidazo[1,2-a]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MDL Number: MFCD11976872
CAS Number: 1260797-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR