3-Chloropyridazine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-1,2-diazine hydrochloride

A135523-1G A135523-5G
77027-290EA 16 USD
77027-290 77027-292
3-Chloropyridazine hydrochloride 95%
3-Chloropyridazine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₃ClN₂·ClH
MDL Number: MFCD12547058
CAS Number: 856847-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR