5-Hydroxy-1H-imidazole-4-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: Bredinin aglycone

A325034-100MG A325034-1G A325034-250MG
76978-136EA 37 USD
76978-136 76978-138 76978-140
5-Hydroxy-1H-imidazole-4-carboxamide 98%
5-Hydroxy-1H-imidazole-4-carboxamide

Precaución: for Research Use Only

Formula: C4H5N3O2
MW: 127.1014 g/mol
MDL Number: MFCD09753122
CAS Number: 56973-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR