4-[(4-Iodobenzyl)oxy]piperidine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 4-((4-Iodobenzyl)oxy)piperidine hydrochloride

A224679-1G A224679-250MG
76975-940EA 72 USD
76975-940 76975-942
4-[(4-Iodobenzyl)oxy]piperidine hydrochloride 95%
4-[(4-Iodobenzyl)oxy]piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₇ClINO
MW: 353.63 g/mol
MDL Number: MFCD13561219
CAS Number: 1220034-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR