3-Ethoxypyridazine 97%

Supplier: Ambeed
Warning

A241108-100G A241108-10G A241108-1G A241108-5G A241108-25G
76976-466EA 553 USD
76976-466 76976-468 76976-470 76976-472 77672-156
3-Ethoxypyridazine 97%
3-Ethoxypyridazine

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.1400 g/mol
MDL Number: MFCD14585053
CAS Number: 62567-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR