5-Bromo-1-isopropylpyridin-2(1H)-one ≥97%
Supplier: AOBChem USA
|
Warning
|
23738-1G
23738-25G
23738-5G
76928-160EA
91.24
USD
76928-160
76928-162
76928-164
5-Bromo-1-isopropylpyridin-2(1H)-one ≥97%
5-Bromo-1-isopropylpyridin-2(1H)-one
Formula:
C₈H₁₀BrNO Storage Temperature: Refrigerator |
MDL Number:
MFCD10699719 CAS Number: 851087-08-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...