[3-Methoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol ≥97% research grade

Supplier: AOBChem USA

Warning

43638-1G 43638-500MG 43638-5G
76889-890EA 471.04 USD
76889-890 76889-892 76889-894
[3-Methoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol ≥97% research grade
[3-Methoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
Formula: C₁₄H₂₁BO₄
Storage Temperature: Ambient
MDL Number: MFCD27936406
CAS Number: 1392421-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR