5-Bromo-4-fluoro-2-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

14863-25G 14863-5G
76925-304EA 233.4 USD
76925-304 76925-306
5-Bromo-4-fluoro-2-iodotoluene ≥97%
5-Bromo-4-fluoro-2-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
Storage Temperature: Ambient
MDL Number: MFCD09800686
CAS Number: 861928-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR