5-Bromo-1-isopropylpyridin-2(1H)-one 98%

Supplier: Ambeed
Warning

A733966-100MG A733966-1G
76879-898EA 30 USD
76879-898 76879-900
5-Bromo-1-isopropylpyridin-2(1H)-one 98%
5-Bromo-1-isopropylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MDL Number: MFCD10699719
CAS Number: 851087-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR