5-Bromo-2-isopropylpyridine 95%

Supplier: Ambeed
Warning

A104148-250MG A104148-5G A104148-1G
76869-304EA 64 USD
76869-304 76869-306 77565-812
5-Bromo-2-isopropylpyridine 95%
5-Bromo-2-isopropylpyridine

Precaución: For research use only.

Formula: C₈H₁₀BrN
MDL Number: MFCD12033382
CAS Number: 1159820-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR