Valeric acid hydrazide 98%

Supplier: Ambeed
Danger

Synonyms: Pentanehydrazide

A299671-250MG A299671-25G A299671-5G
76787-708EA 9 USD
76787-708 76787-710 76787-712
Valeric acid hydrazide 98%
Valeric acid hydrazide

Precaución: for Research Use Only

Formula: C₅H₁₂N₂O
MDL Number: MFCD00025137
CAS Number: 38291-82-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR