Triisopropylsilane 98%

Supplier: Ambeed
Danger

A187865-10G A187865-25G A187865-5G
76784-824EA 11 USD
76784-824 76784-826 76784-828
Triisopropylsilane 98%
Triisopropylsilane

Precaución: for Research Use Only

Formula: C₉H₂₂Si
MDL Number: MFCD00009657
CAS Number: 6485-79-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR