Tetrabutylammonium acetate 98% (contains 10-15%AcOH)

Supplier: Ambeed
Warning

A162456-1000G A162456-100G A162456-10G A162456-500G A162456-5G A162456-25G
76783-918EA 105 USD
76783-918 76783-920 76783-922 76783-924 76783-926 77319-910
Tetrabutylammonium acetate 98% (contains 10-15%AcOH)
Tetrabutylammonium acetate

Precaución: for Research Use Only

Formula: C₁₈H₃₉NO₂
MDL Number: MFCD00043208
CAS Number: 10534-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR