tert-Butyl-2-((diphenylmethylene)amino)acetate 98%

Supplier: Ambeed
Warning

Synonyms: N-(Diphenylmethylene)glycine tert-butyl ester, Glycine, N-(diphenylmethylene)-, 1,1-dimethylethyl ester, tert-Butyl 2-((diphenylmethylene)amino)acetate

A189197-10G A189197-1G A189197-25G A189197-5G A189197-100G A189197-500G
76769-826EA 9 USD
76769-826 76769-828 76769-830 76769-832 77600-534 77625-906
tert-Butyl-2-((diphenylmethylene)amino)acetate 98%
tert-Butyl-2-((diphenylmethylene)amino)acetate

Precaución: for Research Use Only

Formula: C₁₉H₂₁NO₂
MDL Number: MFCD00134280
CAS Number: 81477-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR