(R)-(-)-2-Amino-5-phosphonopentanoic acid 95%
Supplier: Ambeed
|
Warning
|
A166104-10MG
A166104-1MG
A166104-5MG
76690-036EA
104
USD
76690-036
76690-038
76690-040
(R)-(-)-2-Amino-5-phosphonopentanoic acid 95%
(R)-(-)-2-Amino-5-phosphonopentanoic acid
Precaución: for Research Use Only
Formula:
C₅H₁₂NO₅P |
MDL Number:
MFCD00078839 CAS Number: 79055-68-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...