1-Bromononane 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Nonyl bromide
A1471058-100G
A1471058-500G
A1471058-25G
A1471058-1000G
76662-756EA
12
USD
76662-756
76662-758
77592-750
77597-394
1-Bromononane 99%
1-Bromononane
Precaución: for Research Use Only
Formula:
BrH₂C(CH₂)₇CH₃ |
MDL Number:
MFCD00000278 CAS Number: 693-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...