(S)-Isopropyl 2-amino-3-hydroxypropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Ser-OIsoPropyl·HCl

A802373-250MG A802373-5G
76653-458EA 11 USD
76653-458 77784-089
(S)-Isopropyl 2-amino-3-hydroxypropanoate hydrochloride 97%
(S)-Isopropyl 2-amino-3-hydroxypropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₃
MDL Number: MFCD31630717
CAS Number: 37592-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR