5,7-Dichloro-[1,2,4]triazolo[1,5-a]pyridine 97%

Supplier: Ambeed
Warning

A598963-100MG A598963-250MG A598963-1G
76650-046EA 45 USD
76650-046 76650-048 77783-490
5,7-Dichloro-[1,2,4]triazolo[1,5-a]pyridine 97%
5,7-Dichloro-[1,2,4]triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂N₃
MDL Number: MFCD31567139
CAS Number: 2009345-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR