1-Chloroimidazo[1,5-a]pyridine 95+%

Supplier: Ambeed
Warning

A158845-100MG A158845-1G A158845-250MG
76647-124EA 158.08 USD
76647-124 76647-126 76647-128
1-Chloroimidazo[1,5-a]pyridine 95+%
1-Chloroimidazo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MDL Number: MFCD30067671
CAS Number: 1890909-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR