2-((2-(Cyclohexyl(methyl)amino)-2-oxoethyl)thio)acetic acid 95%

Supplier: Ambeed
Warning

A1010575-1G A1010575-100MG A1010575-250MG
76646-714EA 887 USD
76646-714 77782-832 77782-833
2-((2-(Cyclohexyl(methyl)amino)-2-oxoethyl)thio)acetic acid 95%
2-((2-(Cyclohexyl(methyl)amino)-2-oxoethyl)thio)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃S
MDL Number: MFCD06356453
CAS Number: 790232-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR