5-(Hydroxymethyl)resorcinol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dihydroxybenzyl alcohol

34406-0050 34406-0250
TS34406-0050EA 90 USD
TS34406-0050 TS34406-0250
5-(Hydroxymethyl)resorcinol 98%
5-(Hydroxymethyl)resorcinol
Formula: C₇H₈O₃
MW: 140.14 g/mol
Melting Pt: 181…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00016867
CAS Number: 29654-55-5
EINECS: 249-751-8

Order Now

Specification Test Results

Appearance Off-white to beige-pink to brown Crystalline powder
Infrared spectrum Conforms
Melting point 181 to 186 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR