(±)-Taxifolin ≥90% (by HPLC), Sigma-Aldrich®

Warning


M-Clarity™ quality level = MQ100

T4512-100MG T4512-25MG
220226-983EA 421.15 USD
220226-983 220226-984
(±)-Taxifolin ≥90% (by HPLC), Sigma-Aldrich®
(±)-Taxifolin

Taxifolin or dihydroquercetin is a naturally occurring secondary metabolic compound found in many plants such as onions, milk thistle, Douglas fir bark, and French maritime pine bark.

Formula: C₁₅H₁₂O₇
Storage Temperature: Freezer
MDL Number: MFCD11044454
CAS Number: 24198-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR