Paraformaldehyde 90%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
16965-0010
16965-0025
16965-0100
TS16965-0010EA
92.5
USD
TS16965-0010
TS16965-0025
TS16965-0100
Paraformaldehyde 90%, pure
Paraformaldehyde
Formula:
(CH₂O)n Melting Pt: 120…170 °C (dec.) Flash Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00133991 CAS Number: 30525-89-4 UN: 2213 ADR: 4.1,III Merck Index: 1307096 |
Specification Test Results
Appearance | White Powder or granules or prills |
Infrared spectrum | Conforms |
Assay | 89.5 to 93.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...