Paraformaldehyde 96%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
41678-0010
41678-5000
41678-0030
41678-0100
41678-0250
TS41678-0010EA
81.6
USD
TS41678-0010
TS41678-5000
TS41678-0030
TS41678-0100
TS41678-0250
Paraformaldehyde 96%, Extra Pure
Paraformaldehyde
Formula:
(CH₂O)n Melting Pt: 120…170 °C (dec.) Flash Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00133991 CAS Number: 30525-89-4 UN: 2213 ADR: 4.1,III Merck Index: 13,07096 |
Specification Test Results
Appearance | White Powder or beads |
Infrared spectrum | Conforms |
Ash | ≤0.02 % |
pH | 4.0 to 5.5 (10 % suspension) |
Titration with HCl | ≥95 % |
Free acid (HCOOH) | ≤0.03 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...