Magnesium acetate tetrahydrate 98+% ACS

Supplier: Thermo Scientific Chemicals

AC42387-0050 AC42387-5000
TS42387-0050EA 22.3 USD
TS42387-0050 TS42387-5000
Magnesium acetate tetrahydrate 98+% ACS
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
Melting Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration Complexometric 98.0 to 102.0 %
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Barium (Ba) ≤0.001 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤5 ppm
Manganese (Mn) ≤0.001 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR