Benzyldimethyldodecylammonium chloride 98%

Supplier: Ambeed
Warning

A477357-25G A477357-100G
103839-164EA 6 USD
103839-164 76843-100 77004-542
Benzyldimethyldodecylammonium chloride 98%
Benzyldimethyldodecylammonium chloride

Precaución: for Research Use Only

Formula: C₂₁H₃₈ClN
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00137276
CAS Number: 139-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR