Picamilon 98%

Supplier: Ambeed
Warning

A486869-10G A486869-250MG A486869-25G A486869-25MG A486869-50MG A486869-1G A486869-5G
103828-184EA 31 USD
103828-184 103839-258 76695-584 76695-586 76695-588 76695-590 76695-592 77307-568 77307-570
Picamilon 98%
Picamilon

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₃
Boiling Pt: 522.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00186518
CAS Number: 34562-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR