1,2,3,4-Tetrahydroisoquinoline-7-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A172433-100MG A172433-250MG A172433-1G
103806-720EA 218 USD
103806-720 103845-004 76784-294 76784-296 77301-202
1,2,3,4-Tetrahydroisoquinoline-7-carboxylic acid hydrochloride 95%
1,2,3,4-Tetrahydroisoquinoline-7-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO₂
MW: 213.66 g/mol
Boiling Pt: 359.7 °C (760 mm Hg)
Flash Pt: 171.4 °C
Storage Temperature: Ambient
MDL Number: MFCD09056810
CAS Number: 220247-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR