(S)-N-Boc-3-piperidinemethanol 98%

Supplier: Ambeed
Warning

Synonyms: (s)-n-(tert-butoxycarbonyl)-3-hydroxymethylpiperidine, (s)-tert-butyl 3-(hydroxymethyl)tetrahydro-1(2h)-pyridinecarboxylate, (S)-1-BOC-3-(Hydroxymethyl)piperidine, tert-Butyl (S)-3-(hydroxymethyl)piperidine-1-carboxylate

A176134-100G A176134-10G A176134-250MG A176134-25G A176134-1G A176134-5G
103807-118EA 13 USD
103807-118 103845-434 76799-536 76799-538 76799-540 77082-790 77301-424 77301-426
(S)-N-Boc-3-piperidinemethanol 98%
(S)-N-Boc-3-piperidinemethanol

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
Boiling Pt: 308 °C (760 mm Hg)
Flash Pt: 140.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683203
CAS Number: 140695-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR