Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US
Warning


Management of Change (MOC) category = R

BS18-ST BAKRBS18-SA
JTBS18-STCS 104.92 USD
JTBS18-ST JTBS18SA
Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Sodium carbonate
Formula: Na₂CO₃
MDL Number: MFCD00003494
CAS Number: 497-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=49b40b045d7589252effa68b9b7304c684465b5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets B.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Water (H₂O) ≤ 0.5 %
NF - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
EP/BP - Assay (Na₂CO₃) (dried basis) 99.5 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Identification D Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Alkali Hydroxides and Bicarbonates Passes Test
EP/BP - Chloride (Cl) ≤ 125 ppm
EP/BP - Sulfate (SO₄) ≤ 250 ppm
EP/BP - Iron (Fe) ≤ 50 ppm
EP/BP - Loss on Drying ≤ 1.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤ 0.071 %
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Arsenic (As) ≤ 3.1 ppm
JP - Loss on Drying ≤ 0.5 %
JP - Assay (Na₂CO₃) (dried basis) 99.0 - 101.0 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR