2-Propanol ≥99.5%, Laboratory Reagent

Supplier: Honeywell Research Chemicals
Danger

109827-2.5L 109827-4X2.5L
BJ109827-2.5LEA 225.25 USD
BJ109827-2.5L BJ1098274X2.5L
2-Propanol ≥99.5%, Laboratory Reagent
2-Propanol

  • ≥95% for general laboratory use.


Beilstein Number: 635639
RTECS Number: NT8050000
UNSPSC Code: 12191601

Formula: (CH₃)₂CHOH
Boiling Pt: 77…83 °C (1.013 hPa)
Melting Pt: –90 °C
Density: 0.784…0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

IR spectrum complying
Assay (GC) Min. 99.5 %
Non-volatile matter Max. 0.003 %
Water (Karl Fischer) Max. 0.2 %
Autoignition Temperature 750 °F
Vapor Density 2.1 (vs air)
Vapor Pressure 33 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR