DAR-MT ≥98%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
Diaminorhodamine-M triazole
CDX-D0274-M001
CDX-D0274-M005
102989-716EA
351.69
USD
102989-716
102989-718
DAR-MT ≥98%
DAR-MT
Reference compound for the fluorescent (nitric oxide) NO probe DAR-M.
Formula:
C₂₉H₃₁N₅O₃ Storage Temperature: Freezer |
CAS Number:
261351-47-7 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | Dark violet powder. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...