4-Bromo-N-methylpyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

073831-250MG 073831-1G 073831-5G
102533-228EA 640 USD
102533-228 102533-230 102533-232
4-Bromo-N-methylpyrimidin-2-amine ≥95%
4-Bromo-N-methylpyrimidin-2-amine

Hazard: Irritant

Formula: C₅H₆BrN₃
MDL Number: MFCD13193672
CAS Number: 1209458-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR