5-Hydroxy-1H-imidazole-4-carboxamide ≥95%
Supplier: Matrix Scientific
Synonyms:
Bredinin aglycone
072895-250MG
072895-1G
102527-316EA
541.54
USD
102527-316
102527-318
5-Hydroxy-1H-imidazole-4-carboxamide ≥95%
5-Hydroxy-1H-imidazole-4-carboxamide
Hazard: Irritant
Formula:
C4H5N3O2 MW: 127.1014 g/mol |
MDL Number:
MFCD09753122 CAS Number: 56973-26-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...