5-(Trifluoromethyl)oxazol-2-amine ≥95%

Supplier: Matrix Scientific

072279-250MG 072279-1G
102524-036EA 869.23 USD
102524-036 102524-038
5-(Trifluoromethyl)oxazol-2-amine ≥95%
5-(Trifluoromethyl)oxazol-2-amine

Hazard: Irritant

Formula: C₄H₃F₃N₂O
MW: 152.0746 g/mol
MDL Number: MFCD09800568
CAS Number: 714972-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR