2,4-Dichlorobenzothiazole ≥95%

Supplier: Matrix Scientific

072090-1G 072090-5G 072090-10G
102523-038EA 427.69 USD
102523-038 102523-040 102523-042
2,4-Dichlorobenzothiazole ≥95%
2,4-Dichlorobenzothiazole

Hazard: Irritant

Formula: C₇H₃Cl₂NS
MDL Number: MFCD00127716
CAS Number: 3622-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR