Anti-IL-2 Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: MQ1-17H12]

Supplier: Biolegend
500333 500334
500333-BLEA 272.7 USD
500333-BL 10761-576
Anti-IL-2 Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: MQ1-17H12]
Antibodies
Brilliant Violet 650™ anti-human IL-2 [MQ1-17H12]; Isotype: Rat IgG2a, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Sooty Mangabey; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-2
Clonality: Monoclonal
Clone: MQ1-17H12
Conjugation: Brilliant Violet® 650
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus;Sooty Mangabey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR