Human Recombinant sCD40 (from E. coli)
Supplier: Peprotech
310-02-10UG 310-02-50UG 310-02-100UG 310-02-250UG 310-02-500UG 310-02-1MG
10780-634EA 136.5 USD
10780-634 10780-636 10780-638 10780-640 10780-642 10780-644
Human Recombinant sCD40 (from E. coli)
Proteins and Peptides
CD40, a member of the TNF receptor superfamily, is a cell surface protein expressed on B cells, dendritic cells, monocytes, thymic epithelial cells and, at low levels, on T cells. Signaling though CD40 plays an important role in the proliferation and differentiation of B cells, and is critical for immunoglobulin (Ig) class switching. The membrane-anchored CD40-Ligand is expressed almost exclusively on activated CD4+ T lymphocytes. Failure to express CD40L leads to “immunodeficiency with hyper-IgM”, a disease characterized by failure to produce IgG, IgA and IgE. The human CD40L gene codes for a 261 amino acid type II transmembrane protein, which contains a 22 amino acid cytoplasmic domain, a 24 amino acid transmembrane domain, and a 215 amino acid extracellular domain. The soluble form of CD40L is an 18 kDa protein comprising the entire TNF homologous region of CD40L and is generated in vivo by an intracellular proteolytic processing of the full length CD40L. Recombinant Human sCD40 ligand is a 16.3 kDa protein containing 149 amino acid residues comprising the receptor binding TNF-like domain of CD40L.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.