Lead(II,IV) oxide ≥97%

Supplier: BeanTown Chemical

125775-1KG 125775-5KG
BT125775-1KGEA 92 USD
BT125775-1KG BT125775-5KG
Lead(II,IV) oxide ≥97%
Lead(II,IV) oxide

CAS: 1314-41-6; EC No: 215-235-6; MDL No: MFCD00078491; RTECS: OG5425000 UN No: UN1479; Haz Class: 5.1; Packing Group: II Powder; Linear Formula: Pb3O4; MW: 685.60


  • RTECS: OG5425000
  • Powder

Formula: Pb₃O₄
MDL Number: MFCD00078491
CAS Number: 1314-41-6
EINECS: 215-235-6
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR