α,α,α-Trifluoro-m-toluoyl chloride ≥98.0%
Supplier: TCI America
Synonyms:
3-(Trifluoromethyl)benzene-1-carbonyl chloride, 3-(Trifluoromethyl)benzoyl chloride
T1335-25G
T1335-5G
TCT1335-25GEA
52
USD
TCT1335-25G
TCT1335-5G
α,α,α-Trifluoro-m-toluoyl chloride ≥98.0%
α,α,α-Trifluoro-m-toluoyl chloride
CAS Number: 2251-65-2
MDL Number: MFCD00000680
Molecular Formula: C8H4ClF3O
Molecular Weight: 208.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 85
Flash Point (°C): 113
Specific Gravity (20/20): 1.41
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-m-tolyl Chloride
Formula:
C₈H₄ClF₃O Boiling Pt: 85 °C Density: 1.41 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00000680 CAS Number: 2251-65-2 UN: 3265 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...